Skip to Content
Merck
All Photos(2)

Key Documents

905119

Sigma-Aldrich

all-cis-Hexafluorocylclohexane

Synonym(s):

all-cis-1,2,3,4,5,6-Hexafluorocyclohexane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$373.00

$373.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$373.00

About This Item

Empirical Formula (Hill Notation):
C6H6F6
CAS Number:
Molecular Weight:
192.10
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$373.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

form

solid

mp

289-290 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

F[C@@H]1[C@@H]([C@@H]([C@@H]([C@@H]([C@@H]1F)F)F)F)F

InChI key

PQJGSPVYVGXTJW-JMVOWJSSSA-N

Application

The incorporation of fluorine in agrochemical and pharmaceutical compounds is of high interest due to the ability to fine-tune the polarity, pKA-value, conformation and metabolic stability. Derived from a cis-selective hydrogenation developed by Glorius and co-workers, this fluorinated compound displays a diverse utility as a building block for chemical synthesis. All-cis-multifluorinated cyclohexanes are of great interest in material chemistry. All-cis-1,2,3,4,5,6-hexafluorocyclohexane, which has been referred to as “Janus-headed” molecule, is one of the most polar compounds known. Liquid crystalline materials are known applications for all-cis-multifluorinated cyclohexanes.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil S Keddie et al.
Nature chemistry, 7(6), 483-488 (2015-05-21)
The highest-energy stereoisomer of 1,2,3,4,5,6-hexafluorocyclohexane, in which all of the fluorines are 'up', is prepared in a 12-step protocol. The molecule adopts a classic chair conformation with alternate C-F bonds aligned triaxially, clustering three highly electronegative fluorine atoms in close
Mario P Wiesenfeldt et al.
Science (New York, N.Y.), 357(6354), 908-912 (2017-08-12)
All-cis-multifluorinated cycloalkanes exhibit intriguing electronic properties. In particular, they display extremely high dipole moments perpendicular to the aliphatic ring, making them highly desired motifs in material science. Very few such motifs have been prepared, as their syntheses require multistep sequences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service