Skip to Content
Merck
All Photos(2)

Key Documents

D102202

Sigma-Aldrich

1,4-Difluorobenzene

≥99%

Synonym(s):

2,5-Difluorobenzene, p-Difluorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$99.50
100 G
$202.00

$99.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$99.50
100 G
$202.00

About This Item

Empirical Formula (Hill Notation):
C6H4F2
CAS Number:
Molecular Weight:
114.09
Beilstein:
1904541
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$99.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

88-89 °C (lit.)

mp

−13 °C (lit.)

density

1.11 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(F)cc1

InChI

1S/C6H4F2/c7-5-1-2-6(8)4-3-5/h1-4H

InChI key

QUGUFLJIAFISSW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Organic Semiconductor Synthesis: Synthesis of 6,13-difluoropentacene using 1,4-Difluorobenzene showcases its role in developing high-performance organic semiconductors for electronic devices. This research highlights the compound′s utility in enhancing the efficiency and stability of electronic materials (Tripp and Koert, 2020).
  • Hydrophobicity in Chemical Structures: Research on the progressive hydrophobicity of fluorobenzenes, including 1,4-Difluorobenzene, aids in understanding their interactions in biological and chemical systems, which is crucial for the design of hydrophobic surfaces and materials (Kumar et al., 2019).
  • Molecular Coordination Chemistry: Study on the coordination of arenes and phosphines by charge-separated alkaline earth cations using 1,4-Difluorobenzene, which is pivotal for the development of new coordination compounds with potential applications in catalysis and materials science (Garcia et al., 2018).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service