Skip to Content
Merck
All Photos(1)

Key Documents

CF0012

Sigma-Aldrich

Alcohol C2F5-Togni reagent

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$296.00

$296.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$296.00

About This Item

Linear Formula:
C11H10F5IO
CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.06

$296.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

reaction suitability

reaction type: C-C Bond Formation

storage temp.

−20°C

SMILES string

[I]1(OC(c2c1cccc2)(C)C)C(F)(F)C(F)(F)F

InChI

1S/C11H10F5IO/c1-9(2)7-5-3-4-6-8(7)17(18-9)11(15,16)10(12,13)14/h3-6H,1-2H3

InChI key

LCYUDGXZWNAFLM-UHFFFAOYSA-N

Application

Togni-C2F5-reagent is a perfluoroethyl extension of the established Togni-CF3-reagent (754218) exhibiting the same reactivity.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Product of CF Plus Chemicals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluoroalkylation toolbox expands with various reagents for late-stage fluoroalkylation in organic synthesis and medicinal chemistry.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service