Skip to Content
Merck
All Photos(2)

Key Documents

901759

Sigma-Aldrich

AquaMet

Synonym(s):

(4-((4-Ethyl-4-methylpiperazin-1-ium-1-yl)methyl)-1,3-dimesitylimidazolidin-2-ylidene)dichloro(2-isopropoxybenzylidene)ruthenium(II) chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$162.00

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$162.00

About This Item

Empirical Formula (Hill Notation):
C39H55Cl3N4ORu
CAS Number:
Molecular Weight:
803.31
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

$162.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

Application

AquaMet is a highly active catalyst for olefin metathesis in water. The catalyst can be used in metathesis of water-insoluble substrates in ′′classical′′ organic solvents and in reactions in ionic liquids.

Legal Information

Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com. PCT/EP2013/053967

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Highly active catalysts for olefin metathesis in water.
Skowerski K, et al.
Catalysis Science & Technology, 2(12), 2424-2427 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service