Skip to Content
Merck
All Photos(2)

Key Documents

923125

Sigma-Aldrich

Grubbs Catalyst® M801

Umicore

Synonym(s):

Bis[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(benzylidene)ruthenium(II)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$150.00
1 G
$601.00

$150.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$150.00
1 G
$601.00

About This Item

Empirical Formula (Hill Notation):
C49H58Cl2N4Ru
CAS Number:
Molecular Weight:
874.99
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

$150.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

mp

>200 °C

SMILES string

Cl[Ru](c1n(C2=C(C)C=C(C)C=C2C)CCn1C3=C(C)C=C(C)C=C3C)(Cl)(c4n(C5=C(C)C=C(C)C=C5C)CCn4C6=C(C)C=C(C)C=C6C)=CC7=CC=CC=C7

Application

Grubbs Catalyst® M801 is a highly latent metathesis catalysts that can be mixed with reactive monomers and not initiate at ambient temperature.

Learn more about our metathesis catalysts

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

For any further information on product please refer to your local Umicore PMC contact at http://www.pmc.umicore.com
Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service