Skip to Content
Merck
All Photos(1)

Key Documents

88296

Millipore

Thayer Martin Agar (Base)

suitable for microbiology, NutriSelect® Plus

Synonym(s):

GC Agar (Base)

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$167.00

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$167.00

About This Item

UNSPSC Code:
41171611
NACRES:
NA.74

$167.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 13 g/L
peptone, special, 23 g/L
sodium chloride, 5 g/L
starch (soluble), 1 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages

microbiology

suitability

selective for Neisseria spp.

Application

For the selective isolation of Neisseria gonorrhoeae and Neisseria meningitidis from clinical material according to Thayer and Martin. Suitable for transport of inocculated material.

Preparation Note

Dissolve 21.0 g in 240 ml distilled water and autoclave at 121°C for 15 minutes. Add the rehydrated contents of one bottle of Thayer-Martin Supplement I (Cat. No. 44051) and Thayer-Martin Supplement II (Cat. No. 52083) to the medium at approx. 50°C. Additionally add 250 ml of sterile 2% hemoglobin (Cat. No. 08449) solution. Mix gently and pour into sterile petri plates or tubes for slants.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.D. Thayer et al.
Publ. Health Rep., 81, 559-559 (1966)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service