Skip to Content
Merck
All Photos(1)

Key Documents

1.09204

Sigma-Aldrich

Giemsa′s azur eosin methylene blue solution

Giemsa′s azur eosin methylene blue solution

for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.05
Pricing and availability is not currently available.

grade

for microscopy

Quality Level

form

liquid

autoignition temp.

455 °C

composition

C.I. 45380, 2.4 g/L
C.I. 52015 + Azure, 4.1 g/L
contains CH3OH

expl. lim.

5.5-44 % (v/v) Methanol)

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

6.1-7.0 (20 °C in H2O, undiluted)

bp

>65 °C/1013 hPa

transition temp

flash point 18 °C

density

0.99 g/cm3 at 20 °C

suitability

suitable for microscopy

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Giemsa′s Azure Eosin Methylene Blue solution - for microscopy, can be used for the staining of blood and bone marrow smears, paraffin sections and clinical-cytological specimens. Giemsa staining is one of the standard procedures in histology, demonstrating various cells with their morphological features better than with H&E stain. When used in hematological applications, Giemsa′s stain is frequently used in combination with other staining solutions, e. g. with May-Grünwald′s solution for Pappenheim (MGG) overview staining. This ready-to-use staining solution generally stains the nuclei red, based on the molecular interaction between the eosin Y dye and an Azure B-DNA complex. Both dyes assemble to an Eosin Y - Azure B-DNA complex and the intensity of the resulting stain depends on the content of Azure B and the ratio of Azure B : Eosin Y. Furthermore, the resulting stain can vary depending on the influence of fixation, staining times, pH-value of the solutions or buffer substances. In histology and clinico-cytological applications, Giemsa′s staining without additional stainings is used as an extended overview staining method. In this method, the color of the various cell components is influenced by pretreatment of the specimen material. Here, chromatin-containing structures (e. g. cell nuclei) appear in various blue shades, while the acidophilic components show up in a variety of red shades. The 500 ml bottle is sufficient for 3500-5000 stainings. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

For microscopy. For the staining of blood and bone-marrow smears, paraffin sections and clinical-cytological specimens

Analysis Note

For staining results, please refer to the Instructions for Use link under the Documents Section below.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dora Posfai et al.
Cell chemical biology, 27(6), 719-727 (2020-04-25)
Plasmodium vivax infects hepatocytes to form schizonts that cause blood infection, or dormant hypnozoites that can persist for months in the liver before leading to relapsing blood infections. The molecular processes that drive P. vivax schizont and hypnozoite survival remain

Questions

  1. Can product 1092040500 be used as a malarial stain? The specified pH range for this product is 6.0 - 8.2 (H₂O, 20 °C) (undiluted). How can the pH be adjusted to 7.2?

    1 answer
    1. Product 109204 does not have specific claims for staining malarial parasites. The Instructions For Use mention staining Helicobacter pylori, but not malaria, as the product is approved for In Vitro Diagnostic Use. The Giemsa solution 109204 is an unbuffered solution. The instructions advise using buffers at pH 6.4, 6.8, or 7.2, depending on customer preference. The resulting colors obtained with the Giemsa solution largely depend on its pH.

      Helpful?

Reviews

Active Filters

  1. Germany
    • Review 1
    • Vote 1
    5 out of 5 stars.

    Great staining

    Staining was consistently good and as crisp as expected.

    Translate with Google

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service