Skip to Content
Merck
All Photos(1)

Key Documents

88103

Supelco

Tetrapropylammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$441.00

$441.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$441.00

About This Item

Linear Formula:
(CH3CH2CH2)4N(Br)
CAS Number:
Molecular Weight:
266.26
Beilstein:
3567846
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

$441.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

cationic

Quality Level

Assay

≥99.0% (AT)

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 240 nm Amax: 0.04
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].CCC[N+](CCC)(CCC)CCC

InChI

1S/C12H28N.BrH/c1-5-9-13(10-6-2,11-7-3)12-8-4;/h5-12H2,1-4H3;1H/q+1;/p-1

InChI key

BGQMOFGZRJUORO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

The ion-pairing reagents, when utilized as mobile phase additives, enable the elution of ionic and highly polar analytes on reversed phase HPLC columns. The purity of mobile phase additives is quintessential for their successful use. Sigma-Aldrich has a large collection of tailor-made reagents for anionic (quaternary ammonium and phosphonium salts) and cationic (alkanesulfonates) determinations. All mobile phase additives are rigorously examined with special focus on the necessities of modern reversed phase HPLC:

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarika Goel et al.
Journal of the American Chemical Society, 136(43), 15280-15290 (2014-10-15)
The encapsulation of metal clusters (Pt, Ru, Rh) within MFI was achieved by exchanging cationic metal precursors into a parent zeolite (BEA, FAU), reducing them with H2 to form metal clusters, and transforming these zeolites into daughter structures of higher
Peng Bai et al.
Nature communications, 6, 5912-5912 (2015-01-22)
Zeolites play numerous important roles in modern petroleum refineries and have the potential to advance the production of fuels and chemical feedstocks from renewable resources. The performance of a zeolite as separation medium and catalyst depends on its framework structure.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service