Merck
  • Discovery of optimal zeolites for challenging separations and chemical transformations using predictive materials modeling.
Discovery of optimal zeolites for challenging separations and chemical transformations using predictive materials modeling.
Nature communications (2015-01-22)
Peng Bai, Mi Young Jeon, Limin Ren, Chris Knight, Michael W Deem, Michael Tsapatsis, J Ilja Siepmann
ABSTRACT
Zeolites play numerous important roles in modern petroleum refineries and have the potential to advance the production of fuels and chemical feedstocks from renewable resources. The performance of a zeolite as separation medium and catalyst depends on its framework structure. To date, 213 framework types have been synthesized and >330,000 thermodynamically accessible zeolite structures have been predicted. Hence, identification of optimal zeolites for a given application from the large pool of candidate structures is attractive for accelerating the pace of materials discovery. Here we identify, through a large-scale, multi-step computational screening process, promising zeolite structures for two energy-related applications: the purification of ethanol from fermentation broths and the hydroisomerization of alkanes with 18-30 carbon atoms encountered in petroleum refining. These results demonstrate that predictive modelling and data-driven science can now be applied to solve some of the most challenging separation problems involving highly non-ideal mixtures and highly articulated compounds.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Propylamine, ≥99%
Sigma-Aldrich
Propylamine, 98%
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrogen fluoride pyridine, pyridine ~30 %, hydrogen fluoride ~70 %
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Hydrofluoric acid, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrofluoric acid, ACS reagent, 48%
Sigma-Aldrich
Ammonium fluoride, ACS reagent, ≥98.0%
Supelco
Propylamine, analytical standard
Supelco
Tetrapropylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Tetrapropylammonium bromide, 98%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard