Skip to Content
Merck
All Photos(1)

Key Documents

80589

Supelco

Brevetoxin 2 solution

20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Brevetoxin B solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
$429.00

$429.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
0.5 ML
$429.00

About This Item

Empirical Formula (Hill Notation):
C50H70O14
CAS Number:
Molecular Weight:
895.08
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$429.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

form

solution

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in acetonitrile (nominal concentration)

format

single component solution

shipped in

dry ice

storage temp.

−20°C

SMILES string

O1[C@@]2([C@H](O[C@@H](C[C@@H]2O)CC(=C)C=O)C[C@@H]3O[C@H]4[C@@H](O[C@H]5[C@@](O[C@H]6[C@@](O[C@]7([C@@H](O[C@H]8[C@@H](O[C@H]9[C@@H](O[C@H]%10[C@@](O[C@H]%11[C@@H](OC(=O)C=C%11C)C%10)(C9)C)C[C@H]8C)CC7)C6)C)(C5)C)(C\C=C/4)C)C[C@H]31)C

InChI

1S/C50H70O14/c1-25(24-51)14-28-17-37(52)50(8)41(54-28)19-33-34(61-50)18-32-29(55-33)10-9-12-46(4)42(58-32)23-49(7)40(62-46)21-39-47(5,64-49)13-11-30-44(60-39)26(2)15-31-36(56-30)22-48(6)38(57-31)20-35-45(63-48)27(3)16-43(53)59-35/h9-10,16,24,26,28-42,44-45,52H,1,11-15,17-23H2,2-8H3/b10-9-/t26-,28-,29-,30+,31+,32+,33+,34-,35+,36-,37+,38-,39+,40-,41-,42-,44-,45-,46+,47-,48+,49+,50+/m1/s1

InChI key

LYTCVQQGCSNFJU-FGRVLNGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI.
Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

For the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service