Skip to Content
Merck
All Photos(1)

Documents

78688

Supelco

S-Desoxybrevetoxin B2 solution

20 μg/g in methanol (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

42-[[(2R)-2-Amino-2-carboxyethyl]thio]-42-deoxo-41,43-dihydro-43-hydroxybrevetoxin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H79NO16S
CAS Number:
Molecular Weight:
1018.26
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in methanol (nominal concentration)

format

single component solution

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Brevetoxin for the analysis of marine toxins in seafood.

Check out our complete range of marine toxins reference materials

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service