Skip to Content
Merck
All Photos(1)

Key Documents

77450

Supelco

2-Bromoacetophenone

for GC derivatization, LiChropur, ≥99.0%

Synonym(s):

ω-Bromoacetophenone, Phenacyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$186.00

$186.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$186.00

About This Item

Linear Formula:
C6H5COCH2Br
CAS Number:
Molecular Weight:
199.04
Beilstein:
606474
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

$186.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥99.0% (GC)
≥99.0%

form

crystals

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

bp

135 °C/18 mmHg (lit.)

mp

48-51 °C (lit.)
49-50 °C

storage temp.

2-8°C

SMILES string

BrCC(=O)c1ccccc1

InChI

1S/C8H7BrO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H,6H2

InChI key

LIGACIXOYTUXAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Bromoacetophenone is a commonly used derivatization agent for fatty acid detection in biological samples.[1]

Application

Preparation of crystalline esters from acids

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

High-performance liquid chromatography of fatty acids in biological samples.
Lima ES and Abdalla DSP
Analytica Chimica Acta, 465(1-2), 81-91 (2002)
Anabha E Raveendran et al.
Organic & biomolecular chemistry, 8(4), 901-905 (2010-02-06)
Nucleophilic heterocyclic carbenes (NHCs) have been used for the first time as catalysts in the cyclopropanation of ethyl cyanocinnamates with phenacyl bromide by Michael-initiated ring-closure (MIRC).
S Ramalingam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 78(2), 826-834 (2011-01-11)
In this work, the experimental and theoretical spectra of 4-chloro-2-bromoacetophenone (4C2BAP) are studied. FT-IR and FT-Raman spectra of title molecule have been recorded in the region 4000-100 cm(-1). The structural and spectroscopic data of the molecule in the ground state
P A Wender et al.
Organic letters, 1(13), 2117-2120 (2000-06-03)
[formula: see text] 4'-Bromoacetophenone derivatives which upon excitation can generate monophenyl radicals capable of hydrogen atom abstraction were investigated as photoinducible DNA cleaving agents. Pyrrolecarboxamide-conjugated 4'-bromoacetophenones were synthesized, and their DNA cleaving activities and sequence selectivities were determined.
A Kanstrup et al.
Archives of biochemistry and biophysics, 304(2), 332-337 (1993-08-01)
A series of 18 phenacyl bromide and iodoacetamide analogues have been synthesized and used to alkylate Met-398 situated in the S'1 binding site of carboxypeptidase Y. The course of the reactions was monitored by measurements of the peptidase and esterase

Questions

1–2 of 2 Questions  
  1. Do you have references on the use of 2-Bromoacetophenone (also known as phenacyl bromide) Products 115835 and 77450 for making phenacyl esters of carboxylic acids for HPLC analysis?

    1 answer
    1. For information on the use of phenacyl bromide to derivatize carboxylic acids (e.g., fatty acids) for HPLC analysis, please see one or more of the following three references:1. K.J. Longmuir, et al., Determination of monoenoic fatty acid double bond position by permanganate-periodate oxidation followed by high-performance liquid chromatography of carboxylic acid phenacyl esters. Analytical Biochemistry, 167(2), 213-221 (1987).2. T. Hanis et al.,  Determination of fatty acids as phenacyl esters in rat adipose tissue and blood vessel walls by high-performance liquid chromatography. Journal of Chromatography, 452, 443-457 (1988). 3. A. Mehta et al., Rapid quantitation of free fatty acids in human plasma by high-performance liquid chromatography. Journal of Chromatography B Biomedical Science Applications, 719(1-2), 9-23 (1998).

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service