Skip to Content
Merck
All Photos(2)

Key Documents

105120

Sigma-Aldrich

2-Bromo-2′-acetonaphthone

99%

Synonym(s):

Bromomethyl 2-naphthyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$71.60

$71.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$71.60

About This Item

Linear Formula:
C10H7COCH2Br
CAS Number:
Molecular Weight:
249.10
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

82-84 °C (lit.)

solubility

acetonitrile: soluble

functional group

bromo
ketone

SMILES string

BrCC(=O)c1ccc2ccccc2c1

InChI

1S/C12H9BrO/c13-8-12(14)11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2

InChI key

YHXHHGDUANVQHE-UHFFFAOYSA-N

Gene Information

human ... PTPN6(5777)

Related Categories

General description

2-Bromo-2′-acetonaphthone is used for derivatization of captopril to develop a fast and sensitive high-performance liquid chromatographic method for the determination of captopril in plasma[1].2-Bromo-2′-acetonaphthone converts tetra-acids extracted from calcium naphthenate deposits to 2-naphthacyl derivatives[2].

Application

2-Bromo-2′-acetonaphthone was used to determine the concentration of tetra-acids in calcium naphthenate deposits[2]. 2-Bromo-2′-acetonaphthone was used as derivation reagent for determination of fatty acids extracted from Botryococcus braunii by HPLC[3].

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sébastien Simon et al.
Journal of chromatography. A, 1200(2), 136-143 (2008-06-17)
A method is described which allows to determine the content of the so-called C(80) tetra-acid molecules (TA) in calcium naphthenate deposits. The method consists of four steps. Molecules present in the deposit are dissolved in a mixture of toluene and
Jia-You Fang et al.
International journal of pharmaceutics, 276(1-2), 163-173 (2004-04-29)
To improve the drug absorption into and across the skin, fatty acids extracted from Botryococcus braunii were evaluated using in vitro and in vivo techniques with Wistar rats as the animal model. Palmitic acid (C16:0), oleic acid (C18:1), linoleic acid
M Amini et al.
Pharmaceutica acta Helvetiae, 73(6), 303-306 (1999-08-12)
A rapid, simple and sensitive high-performance liquid chromatographic method for the determination of captopril in plasma has been developed. Captopril is derivatized with a new reagent, 2-bromo-2'-acetonaphthone to form a product that showed ultraviolet-absorbing properties. For plasma samples, the protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service