Skip to Content
Merck
All Photos(1)

Key Documents

71715

Supelco

Sodium ionophore I - membrane A

Selectophore, function tested

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700

Quality Level

product line

Selectophore

General description

Visit our Sensor Applications portal to learn more.

Components

Composition;
1.0 wt% Sodium Ionophore I (71732)
66.0 wt% Bis(1-butylpentyl) decane-1,10-diyl diglutarate (ETH 469) (30585)
33.0 wt% Poly(vinyl chloride) high molecular weight (81392)

Other Notes

Ion selective polymer membrane for assaying sodium in undiluted serum or urine with ISE[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Anker et al.
Clinical chemistry, 29(8), 1508-1512 (1983-08-01)
We present an improved Na+-selective liquid membrane electrode for measurement of Na+ concentrations in both undiluted serum and urine. The values for urinary Na+ obtained with the ion-selective electrode agree well with those obtained with the flame photometer. The correlation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service