Skip to Content
Merck
All Photos(1)

Key Documents

68542

Supelco

Perfluoropentanoic acid

analytical standard

Synonym(s):

Nonafluorovaleric acid, PFAS, Nonafluoropentanoic acid, Perfluoropentanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$88.40

$88.40

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$88.40

About This Item

Linear Formula:
CF3(CF2)3COOH
CAS Number:
Molecular Weight:
264.05
Beilstein:
1800087
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$88.40

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 8327
suitable for EPA ACB B21-02
suitable for EPA ACB B23-05b
suitable for EPA OTM-45
suitable for ISO 21675 2019

reg. compliance

suitable for FDA C-010.02

Assay

≥98.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.294 (lit.)

bp

140 °C (lit.)

density

1.713 g/mL at 25 °C (lit.)

application(s)

PFAS testing

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C5HF9O2/c6-2(7,1(15)16)3(8,9)4(10,11)5(12,13)14/h(H,15,16)

InChI key

CXZGQIAOTKWCDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Perfluoropentanoic acid is a persistent organic pollutant belonging to the perfluorinated (PFCs) class of compounds. These compounds are used in a variety of industrial applications, such as textile paints, polishes, adhesives, electronics, waxes, stain repellents, and food packaging.[1][2]

Application

Perfluoropentanoic acid may be used as an analytical reference standard for the determination of the analyte in fish using liquid chromatography–quadrupole-linear ion trap mass spectrometry (LC–QqLIT–MS)[1] and in natural water samples using high-performance liquid chromatography with fluorescence detection (HPLC-FLD).[2]

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a pressurized liquid extraction liquid chromatography?tandem mass spectrometry method for perfluorinated compounds determination in fish
Llorca M, et al.
Journal of Chromatography A, 1216(43), 7195-7204 (2009)
HPLC determination of perfluorinated carboxylic acids with fluorescence detection
Pobozy E, et al.
Microchimica Acta, 172(3-4), 409-417 (2011)

Articles

Liquid Chromatography–Mass Spectrometry (LC-MS) separation of 33 PFAS (Per- and poly-fluoroalkyl substances) using a superficially porous silica particles (SPP) based Ascentis® Express C18 column.

Discover an LC-MS/MS method for the analysis of 16 per- and polyfluoroalkyl compounds (PFAS) in milk using QuEChERS extraction technique following FDA C-010.02 guidelines.

Learn how method EPA 533 is used for sample collection and preparation, featuring Supelclean™ ENVI-WAX™ SPE cartridges for LC-MS analysis on an Ascentis® Express PFAS HPLC Column.

Find more about the analysis of 16 PFAS in salmon via QuEChERS extraction and LC-MS/MS characterization following FDA Method C-010.02.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service