Perfluoropentanoic acid
Synonym(s):
Nonafluorovaleric acid, Nonafluoropentanoic acid, Perfluoropentanoic acid
Linear Formula:
CF3(CF2)3COOH
CAS Number:
Molecular Weight:
264.05
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3