Skip to Content
Merck
All Photos(1)

Documents

61696

Supelco

Heptaldehyde

analytical standard

Synonym(s):

1-Heptanal, Aldehyde C7, Enanthaldehyde, Oenanthaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CHO
CAS Number:
Molecular Weight:
114.19
Beilstein:
1560236
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.413 (lit.)

bp

153 °C (lit.)

mp

−43 °C (lit.)

density

0.821 g/mL at 20 °C
0.817 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCC=O

InChI

1S/C7H14O/c1-2-3-4-5-6-7-8/h7H,2-6H2,1H3

InChI key

FXHGMKSSBGDXIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Li et al.
Clinica chimica acta; international journal of clinical chemistry, 436, 59-67 (2014-05-13)
Breast cancer (BC) remains the most commonly diagnosed malignancy in women. We investigated 4 straight aldehydes in the exhaled breath as potential early BC diagnostic biomarkers. End-tailed breath were collected by Bio-VOC® sampler and assayed by gas chromatography-mass spectrometry. Kruskal-Wallis
Jairo Torres Magalhães-Junior et al.
Analytical and bioanalytical chemistry, 406(26), 6691-6700 (2014-08-31)
Visceral leishmaniasis (VL) is a zoonosis whose etiologic agent in the Americas is Leishmania infantum, and dogs are the main host. Research and innovation in diagnostic techniques are essential to improve the surveillance and control of VL in endemic areas.
Che-Hong Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(10), 3074-3079 (2015-02-26)
Correcting a genetic mutation that leads to a loss of function has been a challenge. One such mutation is in aldehyde dehydrogenase 2 (ALDH2), denoted ALDH2*2. This mutation is present in ∼ 0.6 billion East Asians and results in accumulation
Zahra Hadian et al.
Iranian journal of pharmaceutical research : IJPR, 13(2), 393-404 (2014-09-23)
Omega-3 fatty acids (FAs) have been shown to prevent cardiovascular disease. The most commonly used omega-3 fatty acids like eicosapentaenoic acid (EPA) and docosahexaenoic acid (DHA) are highly vulnerable to oxidation and therefore, have short shelf life. Recent advances in
Matthew W Waugh et al.
Biochemistry, 53(34), 5537-5543 (2014-08-22)
The reaction catalyzed by cyanobacterial aldehyde deformylating oxygenase is of interest both because of its potential application to the production of biofuels and because of the highly unusual nature of the deformylation reaction it catalyzes. Whereas the proton in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service