Skip to Content
Merck
All Photos(2)

Documents

76735

Supelco

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride

for GC derivatization, LiChropur, ≥99.0% (AT)

Synonym(s):

PFBHA·HCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6F5CH2ONH2·HCl
CAS Number:
Molecular Weight:
249.57
Beilstein:
4031190
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥99.0% (AT)

form

solid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

technique(s)

gas chromatography (GC): suitable

mp

212-218 °C
227 °C (subl.) (lit.)

solubility

H2O: 5%, clear

SMILES string

Cl.NOCc1c(F)c(F)c(F)c(F)c1F

InChI

1S/C7H4F5NO.ClH/c8-3-2(1-14-13)4(9)6(11)7(12)5(3)10;/h1,13H2;1H

InChI key

HVMVKNXIMUCYJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride (PFBHA.HCl) is a derivatizing agent.

Application

PFBHA.HCl has been used for derivatization of metabolites with carbonyl groups followed by analysis via GC-MS. It has also been used in aldehyde derivatization followed by aldehydic hydrolysis using GC/MS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Sensitive derivatizing agent for electron capture gas chromatographic analysis of carbonyl-containing compounds: keto steroids and carbohydrates

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Degradation of fluoranthene by Pasteurella sp. IFA and Mycobacterium sp. PYR-1:isolation and identification of metabolites.
Sepic E, Bricelj M, Leskovsek H.
Journal of Applied Microbiology, 85(4), 746-754 null
Y W Lin et al.
Applied occupational and environmental hygiene, 15(11), 855-862 (2000-11-04)
Ketones are the major positive interferences for an aldehyde dynamic air sampler that consists of 200-mg 20 percent (w/w) O-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine hydrochloride (PFBHA) on Tenax TA contained in a Pyrex tube 7-mm OD, 5-mm ID, and 70-mm in length, that utilizes
Giuseppe Pieraccini et al.
Journal of chromatography. A, 955(1), 117-124 (2002-06-14)
Glutaraldehyde is used primarily in hospital environments for the disinfection of various instruments (e.g., endoscopes). We describe in this paper the measurement of glutaraldehyde in a hospital environment using solid-phase microextraction. The method includes, prior to sampling, the adsorption of
R S Spaulding et al.
Analytical and bioanalytical chemistry, 372(7-8), 808-816 (2002-05-16)
The employment of O-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine (PFBHA) derivatization along with bis-(trimethylsilyl)trifluoroacetamide (BSTFA) or N, N-( tert-butyldimethylsilyl)trifluoroacetamide (MTBSTFA) derivatization is a popular method for measurement of oxygenated organics in environmental and biological samples. Most notably, the derivatization method enables the measurement of atmospheric
Chunhui Deng et al.
Rapid communications in mass spectrometry : RCM, 19(5), 647-653 (2005-02-09)
In the current work, a simple, rapid, accurate and inexpensive method was developed for the determination of acetone in human blood. The proposed method is based on derivatization with O-(2,3,4,5,6-pentafluorobenzyl)hydroxylamine hydrochloride (PFBHA), followed by headspace liquid-phase microextraction (HS-LPME) and gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service