Skip to Content
Merck
All Photos(1)

Documents

41782

Millipore

MRS Agar, Vegitone

suitable for microbiology, NutriSelect® Plus

Synonym(s):

DeMan, Rogosa and Sharpe Agar, Vegitone, Vegitone MRS Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
ammonium citrate, 2.0 g/L
bromocresol green, 0.04 g/L
captan, 0.004 g/L
dextrose, 20.0 g/L
dipotassium phosphate, 2.0 g/L
magnesium sulfate, 0.1 g/L
manganese sulfate, 0.05 g/L
2-phenylethyl alcohol, 3.0 g/L
proteose peptone (vegetable), 10.0 g/L
sodium acetate, 5.0 g/L
yeast extract, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.5±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages

microbiology

suitability

selective and differential for Lactobacillus spp.
selective by low pH for Bifidobacterium spp.

Related Categories

General description

This MRS Agar is free of animal derived material. It is recommended for the isolation and cultivation of Lactobacillus species.

Application

MRS Agar, Vegitone has been used for culturing yogurt to analyze lactic acid bacteria (LAB). It has also been used in culturing of Lactobacillus reuteri colonies.

Preparation Note

Suspend 62.2 g in 1000 ml distilled water containing 1 ml polysorbate 80 (TWEEN® 80; Cat. No. P8074). Boil to dissolve the medium completely and mix thoroughly. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. If necessary adjust the pH with glacial acetic acid after sterilization.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

He Ni et al.
Heliyon, 4(10), e00875-e00875 (2018-11-06)
In this study aqueous extracts from salal berry (SB) and blackcurrant pomace (BCP) were used to reformulate yogurt and the anti-diabetic properties of the beverage were investigated during 4 weeks of cold storage at 4 °C. Results indicated that α-amylase
Ferdinand Sedlmayer et al.
Nature communications, 9(1), 1822-1822 (2018-05-10)
Quorum sensing is a promising target for next-generation anti-infectives designed to address evolving bacterial drug resistance. The autoinducer-2 (AI-2) is a key quorum-sensing signal molecule which regulates bacterial group behaviors and is recognized by many Gram-negative and Gram-positive bacteria. Here

Articles

Traditional methods are based morphology, staining methods, enzyme reactions (metabolism) and diverse media.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service