Skip to Content
Merck
All Photos(1)

Key Documents

88517

Millipore

BSM Agar

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Bifidus Selective Agar

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$145.00
500 G
$258.00

$145.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$145.00
500 G
$258.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$145.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

6.8±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

selective and differential for Bifidobacterium spp.

Application

For the selective isolation and identification of Bifidobacteria. The medium is used for quality control in the manufacture of dairy products.

Preparation Note

Dissolve 55.5 g BSM-Agar in 1 litre water. Sterilise by autoclaving at 121°C for 15 min. Cool to 45-50°C and aseptically add 116mg BSM supplement (Cat. No. 83055; freshly dissolved in 1 to 5 ml sterile water). Mix well and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bifidobacteria are important indicators of quality control in the manufacture of dairy products. Bifidobacteria Selective Media (BSM) allows for easy and fast quality control of yoghurt.

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

  1. Hi support, I was wondering whether BSM Agar (NutriSelect Basic) would be suiable for selection of Bifidiobacterium others than resident in dairy products? Specifically, I am interested in isolating Bifidobacterium pullorum from poultry faeces.

    1 answer
    1. Yes, this product is selective and differential for all Bifidobacterium spp. Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/199/251/88517dat.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service