41290
Pelargonin chloride
analytical standard
Synonym(s):
3,5-Bis(glucosyloxy)-4′,7-dihydroxyflavylium chloride, Monardin, Pelargonidin 3,5-di-O-glucoside chloride
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥88% (HPLC)
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
neat
storage temp.
−20°C
SMILES string
[Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)cc(O)cc3[o+]c2-c5ccc(O)cc5)[C@H](O)[C@@H](O)[C@@H]1O
InChI
1S/C27H30O15.ClH/c28-8-17-19(32)21(34)23(36)26(41-17)39-15-6-12(31)5-14-13(15)7-16(25(38-14)10-1-3-11(30)4-2-10)40-27-24(37)22(35)20(33)18(9-29)42-27;/h1-7,17-24,26-29,32-37H,8-9H2,(H-,30,31);1H/t17-,18-,19-,20-,21+,22+,23-,24-,26-,27-;/m1./s1
InChI key
DIRROHKULXIUCB-DHJOXOLYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Pelargonin chloride can be used as standard in the quantitative determination of the total amount of flavonoids like anthocyanin content present in sour cherries using high performance liquid chromatography(HPLC).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service