Skip to Content
Merck
All Photos(1)

Key Documents

379263

Sigma-Aldrich

2-Methyl-2-butene

technical grade, 90%

Synonym(s):

β-Isoamylene, Amylene

Sign Into View Organizational & Contract Pricing

Select a Size

800 ML
$477.00

$477.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
800 ML
$477.00

About This Item

Linear Formula:
CH3CH=C(CH3)2
CAS Number:
Molecular Weight:
70.13
Beilstein:
1361353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$477.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

technical grade

Quality Level

vapor density

2.4 (vs air)

vapor pressure

25.7 psi ( 55 °C)
7.95 psi ( 20 °C)

Assay

90%

form

liquid

expl. lim.

8.7 %

refractive index

n20/D 1.385 (lit.)

bp

35-38 °C (lit.)

mp

−134 °C (lit.)

density

0.662 g/mL at 25 °C (lit.)

SMILES string

C\C=C(\C)C

InChI

1S/C5H10/c1-4-5(2)3/h4H,1-3H3

InChI key

BKOOMYPCSUNDGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-2-butene (β-Isoamylene, Amylene) is a trisubstituted olefin. It acts as guest and forms stable solid host-guest complexes with self-assembled benzophenone bis-urea macrocycles.[1] The effect of active chlorine formulations on photodecomposition of 2-methyl-2-butene has been studied.[2] Its reaction with ozone in gas-phase has been reported to afford carbonym derivatives.[3]

Application

2-Methyl-2-butene may be employed as precursor for peroxyacetyl nitrate for the calibration purposes.[3]

Recommended products

Brass hose adapter Z146811 or brass body mini gas regulator Z513539 is recommended.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F

Flash Point(C)

-20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Gas-phase reaction of ozone with 2-methyl-2-butene: Dicarbonyl formation from Criegee biradicals.
Grosjean D.
Environmental Science & Technology, 24(9), 1428-1432 (1990)
Yunseok Im et al.
The Science of the total environment, 409(13), 2652-2661 (2011-04-29)
Active chlorine comprising hypochlorite (OCl⁻), hypochlorous acid (HOCl) and chlorine (Cl₂) is the active constituent in bleach formulations for a variety of industrial and consumer applications. However, the strong oxidative reactivity of active chlorine can cause adverse effects on both
Michael F Geer et al.
The Journal of organic chemistry, 78(11), 5568-5578 (2013-05-16)
This manuscript investigates how incorporation of benzophenone, a well-known triplet sensitizer, within a bis-urea macrocycle, which self-assembles into a columnar host, influences its photophysical properties and affects the reactivity of bound guest molecules. We further report the generation of a
Charles K Westbrook et al.
The journal of physical chemistry. A, 119(28), 7462-7480 (2015-03-31)
Two experimental studies have been carried out on the oxidation of 2-methyl-2-butene, one measuring ignition delay times behind reflected shock waves in a stainless steel shock tube, and the other measuring fuel, intermediate, and product species mole fractions in a
Yu-Zhe Chen et al.
The Journal of organic chemistry, 70(12), 4676-4681 (2005-06-04)
Photosensitized oxidation of trialkylalkenes 2-methyl-2-pentene (1), 1-methylcyclohexene (2), trans-3-methyl-2-pentene (3), cis-3-methyl-2-pentene (4), and 2-methyl-2-butene (5) included in the internal framework of Na-ZSM-5 zeolites was investigated. The zeolite samples having adsorbed the alkenes were suspended in isooctane, and the sensitizer, tetraphenylporphyrin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service