Skip to Content
Merck
All Photos(2)

Documents

257486

Sigma-Aldrich

2-Methyl-1-butene

98%

Synonym(s):

γ-Isoamylene, 1-Isoamylene, 2-Methyl-1-butylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2C(CH3)=CH2
CAS Number:
Molecular Weight:
70.13
Beilstein:
505975
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

9.98 psi ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.378 (lit.)

bp

31 °C (lit.)

mp

−137 °C (lit.)

density

0.65 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(C)=C

InChI

1S/C5H10/c1-4-5(2)3/h2,4H2,1,3H3

InChI key

MHNNAWXXUZQSNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reactivity of 2-methyl-1-butene in the etherification reaction with ethanol catalyzed by a strongly acidic macroreticular resin catalyst has been investigated. Kinetics and mechanism of addition of hydrogen and deuterium chloride to 2-methyl-1-butene in nitromethane has been investigated.

Application


  • Combustion Properties Analysis: A detailed study on the combustion properties of a gasoline-like blend of pentene isomers, including 2-Methyl-1-butene, provided insights into laminar flame speed, ignition delay time, and CO laser absorption, aiding in the optimization of fuel formulations for enhanced engine performance (Gregoire et al., 2023).

  • Hydrogenation Mechanism Study: Research utilizing the parahydrogen-induced polarization technique explored the mechanisms of methylenecyclobutane hydrogenation over supported metal catalysts, with 2-Methyl-1-butene serving as a key intermediate, offering potential advancements in catalytic process development (Salnikov et al., 2022).

  • Reaction Kinetics with Hydrogen Atoms: An extensive theoretical study examined the reactions of hydrogen atoms with small alkenes, including 2-Methyl-1-butene, shedding light on the hierarchical reaction mechanisms crucial for understanding and designing better chemical synthesis processes (Power et al., 2021).

  • Ozonolysis of Asymmetric Alkenes: A study on the ozonolysis of asymmetric alkenes, including 2-Methyl-1-butene, used matrix isolation and FT-IR spectroscopy to investigate the chemical pathways, providing valuable insights into atmospheric chemistry and pollutant degradation (Wang et al., 2020).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-34.6 °F - closed cup

Flash Point(C)

-37 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etherification rates of 2-methyl-2-butene and 2-methyl-1-butene with ethanol for environmentally clean gasoline production.
Oktar N, et al.
Journal of Chemical Technology and Biotechnology, 74(2), 155-161 (1999)
Kinetics and mechanism of addition of acids to olefins. III. Addition of hydrogen chloride to 2-methyl-1-butene, 2-methyl-2-butene, and isoprene in nitromethane.
Pocker Y, et al.
Journal of the American Chemical Society, 91(15), 4199-4205 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service