Skip to Content
Merck
All Photos(1)

Key Documents

36427

Supelco

Nitrite Nitrogen Standard for IC

certified reference material, 1000 mg/L N as nitrite in water (pH ~11 nominal concentration), TraceCERT®

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$55.00

$55.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$55.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$55.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1000 mg/L N as nitrite in water (pH ~11 nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

storage temp.

2-8°C

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

Nitrite nitrogen standard for IC has been used as working standard in the estimation of nitrite ion present in water via ultraviolet-visible light spectrophotometry.[1]

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L N as nitrite in water (pH ~11), prepared with high purity NaNO2, NaOH and water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water Analysis Using LAMBDA UV-Visible Spectrophotometers: Nitrite Determination null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service