Skip to Content
Merck
All Photos(1)

Key Documents

TKN1000

Supelco

Kjeldahl Nitrogen, Total (TKN) 1000mg/L Calibration Standard

certified reference material

Synonym(s):

TKN Calibration Standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$102.00
500 ML
$146.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$102.00
500 ML
$146.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC TKN1000-100

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is a high-quality wet chemistry standard, produced and certified following ISO 17034 and ISO/IEC 17025 double accreditation—the highest quality system for reference material producers. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

The Total Kjeldahl Nitrogen (TKN) method is used to determine the sum of organic nitrogen, ammonia (NH3) and ammonium (NH4+) contents in the chemical analysis of soil, water and wastewater.

Features and Benefits

  • Provided as 100-mL and 500-mL whole-volume samples
  • Solvent/Matrix: DI Water
  • Traceable to NIST standard reference materials (SRM′s)

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn how water impurities can impact the Kjeldahl nitrogen method. Get recommendations for accurate Kjeldahl analysis and total Kjeldahl nitrogen (TKN) tests. Select the optimal water purification system.

Questions

1–3 of 3 Questions  
  1. Could you provide the user guide for the TKN method? We have a Suplco reflectometer in the lab.does the TKN method work with that? Also, what’s the better approach for assessing organic nitrogen in waste samples: TKN + ammonia or digestive solution + NO3

    1 answer
    1. This product is a calibration standard used to determine the sum of organic nitrogen, ammonia (NH3), and ammonium (NH4+) contents in the chemical analysis of soil, water, and wastewater. However, no user guide for the TKN method is available, and this product does not work with the Supelco reflectometer.
      For assessing organic nitrogen in waste samples, a common approach is to use the TKN method in combination with ammonia measurements. Alternatively, using a digestive solution followed by nitrate (NO3) analysis can also be effective. There are some TKN methods available from external sources. Please check the link: https://www.epa.gov/sites/default/files/2015-08/documents/method_351-2_1993.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service