Skip to Content
Merck
All Photos(2)

Key Documents

33880

Supelco

AOZ-d4

VETRANAL®, analytical standard

Synonym(s):

3-Aminooxazolidin-2-one-d4, 4,4,5,5-Tetradeutero-3-amino-oxazolidin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$682.00

$682.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$682.00

About This Item

Empirical Formula (Hill Notation):
C3H2D4N2O2
CAS Number:
Molecular Weight:
106.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$682.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+4

SMILES string

[2H]C1([2H])OC(=O)N(N)C1([2H])[2H]

InChI

1S/C3H6N2O2/c4-5-1-2-7-3(5)6/h1-2,4H2/i1D2,2D2

InChI key

KYCJNIUHWNJNCT-LNLMKGTHSA-N

General description

It is an isotopically labelled solution of AOZ, also known as 3-amino-2-oxazolidinone, a metabolite of the synthetic nitrofuran veterinary antibiotic― furazolidone.

Application

The analytical standard can be used as an internal standard for the following:
  • Multi-residue analysis of four nitrofuran metabolites residues in 120 fish samples by ultra high-performance liquid chromatography (UHPLC) combined with tandem mass spectrometry (MS/MS)
  • Determination of four nitrofuran metabolites in a variety of food samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS) following their extraction using a solid-phase microextraction (SPME) probe
  • Simultaneous estimation of nitrofuran and chloramphenicol residues in animal-based food products using UHPLC, coupled with high-resolution mass spectrometry (HRMS)
  • Development of a UHPLC-MS/MS-based method for the estimation of nitrofurans along with their metabolites in biological samples
  • Liquid-liquid extraction of chloramphenicol, florfenicol, and nitrofuran metabolites from honey samples for their analysis by LC-MS/MS
  • Estimation of nitrofuran metabolites in poultry muscle and eggs using liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS).[1][2]

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of nitrofuran metabolic residues in salt by liquid chromatography--tandem mass spectrometry
Pereira S.A, et al.
Analytica Chimica Acta, 514(1), 9-13 (2004)
Determination of nitrofuran metabolites in poultry muscle and eggs by liquid chromatography-tandem mass spectrometry
Finzi KJ, et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 824(1-2), 30-35 (2005)
EunChae Ryu et al.
Environmental science and pollution research international, 28(6), 7551-7563 (2020-10-11)
A simplified method is described for reducing the analysis time of nitrofurans (NFs) and nitrofuran metabolites (NFMs) in the aquatic animals. Most existing HPLC-MS/MS methods are intended only for NFMs and are based on their fast metabolic transformations. We optimized

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service