Skip to Content
Merck
All Photos(2)

Key Documents

32512

Supelco

Nitrofurazone-13C,15N2

VETRANAL®, analytical standard

Synonym(s):

5-Nitro-2-furaldehyde semicarbazone-13C,15N2

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,020.00

$1,020.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$1,020.00

About This Item

Empirical Formula (Hill Notation):
13CC5H6O415N2N2
CAS Number:
Molecular Weight:
201.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$1,020.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

mass shift

M+3

SMILES string

N[13C](=O)[15NH]\[15N]=C\c1ccc(o1)[N+]([O-])=O

InChI

1S/C6H6N4O4/c7-6(11)9-8-3-4-1-2-5(14-4)10(12)13/h1-3H,(H3,7,9,11)/b8-3+/i6+1,8+1,9+1

InChI key

IAIWVQXQOWNYOU-LTGGZAIESA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Muta. 2 - Repr. 2 - Skin Sens. 1 - STOT RE 2

Target Organs

male reproductive organs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service