Skip to Content
Merck
All Photos(2)

Key Documents

33349

Supelco

AMOZ

VETRANAL®, analytical standard

Synonym(s):

3-Amino-5-morpholinomethyl-2-oxazolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$871.00

$871.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$871.00

About This Item

Empirical Formula (Hill Notation):
C8H15N3O3
CAS Number:
Molecular Weight:
201.22
Beilstein:
179765
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$871.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

115-120 °C

suitability

passes test for identity

application(s)

clinical testing

format

neat

SMILES string

NN1CC(CN2CCOCC2)OC1=O

InChI

1S/C8H15N3O3/c9-11-6-7(14-8(11)12)5-10-1-3-13-4-2-10/h7H,1-6,9H2

InChI key

TVHAMVOINIHMEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

AMOZ has been used as analytical standard for the determination of residues of nitrofurans and their metabolites, AMOZ (5-methylmorpholino-3-amino-2-oxazolidinone) in animal tissues using liquid chromatography with tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. How can we identify ISO 17034 accredited standards

    1 answer
    1. Products that are ISO 17034 accredited will be explicitly stated on the corresponding product detail page. Please see the link below to review a number of examples:
      https://www.sigmaaldrich.com/search/iso-17034?focus=products&page=1&perpage=30&sort=relevance&term=iso%2017034&type=product

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service