Skip to Content
Merck
All Photos(2)

Key Documents

32508

Supelco

Proquinazid

PESTANAL®, analytical standard

Synonym(s):

6-Iodo-2-propoxy-3-propyl-4(3H)quinazolinone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$260.00

$260.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$260.00

About This Item

Empirical Formula (Hill Notation):
C14H17IN2O2
CAS Number:
Molecular Weight:
372.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$260.00


Estimated to ship onMay 17, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCOC1=Nc2ccc(I)cc2C(=O)N1CCC

InChI

1S/C14H17IN2O2/c1-3-7-17-13(18)11-9-10(15)5-6-12(11)16-14(17)19-8-4-2/h5-6,9H,3-4,7-8H2,1-2H3

InChI key

FLVBXVXXXMLMOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proquinazid belongs to the quinazolinone class of fungicides.[1]

Application

Proquinazid may be used as an analytical reference standard for the determination of the analyte in:
  • Honeybees by dispersive solid-phase extraction (dSPE), followed by liquid and gas chromatography combined with tandem mass spectrometry (LC-MS/MS and GC-MS/MS) operating in electron ionization (EI) and multiple reaction monitoring (MRM) modes of detection.[2]
  • Fruits and vegetables by methanolic extraction, followed by ultra-high performance liquid chromatography (UHPLC) coupled to MS/MS operating in electrospray ionization (ESI) and MRM modes of detection.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of proquinazid on appressorial development of the barley powdery mildew fungus Blumeria graminis f. sp hordei.
Gilbert SR, et al.
Pesticide Biochemistry and Physiology, 94(2-3), 127-132 (2009)
Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry-Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(2-3), 100-114 (2016)
A simple multi-residue method for the determination of pesticides in fruits and vegetables using a methanolic extraction and ultra-high-performance liquid chromatography-tandem mass spectrometry: optimization and extension of scope.
Hanot V, et al.
Journal of Chromatography A, 1384(2-3), 53-66 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service