Skip to Content
Merck
All Photos(2)

Documents

32098

Supelco

Meptyldinocap

PESTANAL®, analytical standard

Synonym(s):

(RS)-2-(1-Methylheptyl)-4,6-dinitrophenyl crotonate, 2-(1-Methylheptyl)-4,6-dinitrophenyl (2E)-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2O6
CAS Number:
Molecular Weight:
364.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCC(C)c1cc(cc(c1OC(=O)\C=C\C)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C18H24N2O6/c1-4-6-7-8-10-13(3)15-11-14(19(22)23)12-16(20(24)25)18(15)26-17(21)9-5-2/h5,9,11-13H,4,6-8,10H2,1-3H3/b9-5+

InChI key

NIOPZPCMRQGZCE-WEVVVXLNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Meptyldinocap is an isomer of dinocap fungicide with reduced environmental impact and mammalian toxicity. Meptyldinocap is a widely used contact fungicide for the control of powdery mildew on grapevines.

Application

Meptyldinocap may be used as a reference standard for the determination of the analyte in various fruit matrices and edible oils by liquid chromatography coupled with tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of pesticides in edible oils by liquid chromatography-tandem mass spectrometry employing new generation materials for dispersive solid phase extraction clean-up.
Dias JV, et al.
Journal of Chromatography A, 1462(6), 8-18 (2016)
Meptyldinocap-a highly active fungicide with curative, eradicant and protectant activity against powdery mildew (Uncinula necator [Erysiphe necator]) on grapevine.
Wittrock A, et al.
Julius-Kuhn Archive (2010)
A fast, inexpensive, and safe method for residue analysis of meptyldinocap in different fruits by liquid chromatography/tandem mass spectrometry.
Banerjee K, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 93(6), 1957-1964 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service