Skip to Content
Merck
All Photos(1)

Key Documents

37909

Supelco

Metconazole

PESTANAL®, analytical standard, mixture of stereoisomers

Synonym(s):

5-(4-Chlorophenylmethyl)-2,2-dimethyl-1-(1H-1,2,4-triazol-1-ylmethyl)cyclopentanol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$166.00

$166.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$166.00

About This Item

Empirical Formula (Hill Notation):
C17H22ClN3O
CAS Number:
Molecular Weight:
319.83
Beilstein:
8333708
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$166.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1(C)CCC(Cc2ccc(Cl)cc2)C1(O)Cn3cncn3

InChI

1S/C17H22ClN3O/c1-16(2)8-7-14(9-13-3-5-15(18)6-4-13)17(16,22)10-21-12-19-11-20-21/h3-6,11-12,14,22H,7-10H2,1-2H3

InChI key

XWPZUHJBOLQNMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metconazole is a lipophilic and a highly active compound which is known for its application as a broad-spectrum triazole fungicide.[1]

Application

Metconazole may be used as a reference standard for the identification of metconazole in:
  • Food and environmental matrices by high-performance liquid chromatography (HPLC) equipped with multiple wavelength ultraviolet (UV) detector.[1]
  • Fruits and vegetables by gas chromatography (GC) equipped with Q Extractive Orbitrap consisting of electron ionization (EI) source and hybrid quadrupole Orbitrap mass spectrometer.[2]
  • Chlorophyll-containing matrix by dispersive-solid phase extraction (d-SPE) and GC with tandem mass spectrometry (MS/MS).[3]
  • Honeybees[4] as well as bee pollen samples[5] by d-SPE followed by analyses using LC-MS/MS and GC-MS/MS and in earthworms by LC-MS/MS with electrospray ionization (ESI).[6]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of a method for the simultaneous determination of multi-class pesticides in earthworms by liquid chromatography coupled to tandem electrospray mass spectrometry.
Daniele G, et al.
Analytical and Bioanalytical Chemistry, 1426, 1-10 (2018)
Enantioselective determination of metconazole in multi matrices by high-performance liquid chromatography.
He R, et al.
Talanta, 178, 980-986 (2018)
Jing Li et al.
Pesticide biochemistry and physiology, 153, 152-160 (2019-02-13)
Validamycin, known to interfere with fungal energy metabolism by inhibiting trehalase, has been extensively used to control plant diseases caused by Rhizoctonia spp. However, the effect of validamycin on controlling Fusarium graminearum has not been previously reported. In this study
Kai Zhang et al.
Journal of chromatography. A, 1519, 100-109 (2017-09-14)
Covalent organic frameworks (COFs), as an emerging class of crystalline porous organic polymers, have great potential for applications in chromatographic separation owning to their fascinating crystalline structures and outstanding properties. However, development of COF materials as novel stationary phases in
Yabing Duan et al.
Plant disease, 103(5), 929-937 (2019-03-19)
Fusarium graminearum species complex (FGSC), causing Fusarium head blight (FHB) of wheat, has species-specific geographical distributions in wheat-growing regions. In recent years, benzimidazole resistance of FHB pathogens has been largely widespread in China. Although the demethylation inhibitor fungicide metconazole has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service