Skip to Content
Merck
All Photos(1)

Key Documents

288608

Supelco

Silica gel Inorganic Sorbent

high-purity grade, 130-270 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$340.00

$340.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$340.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NB.21

$340.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Product Name

Silica gel, high-purity grade, pore size 60 Å, 130-270 mesh, for column chromatography

grade

for column chromatography
high-purity grade

Quality Level

form

powder

technique(s)

LPLC: suitable

surface area

~500 m2/g

matrix

Silica

matrix active group

silica

particle size

130-270 mesh

pore size

0.75 cm3/g pore volume
60 Å

bp

2230 °C

mp

>1600 °C

suitability

suitable for column chromatography

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is a rigid 3D network of colloidal silica.[1] It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.[2] Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction[1]

Application

Silica gel has been used as a column material in the preparation of nanoscale iron oxide−based materials to be used in the catalysis of different hydrogenation reactions, which is monitored using gas chromatography (GC) technique.[3]
Silica gel was used to compare MCM-41 type mesoporous adsorbents.[4]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hydrogenation using iron oxide?based nanocatalysts for the synthesis of amines.
Jagadeesh VR, et al.
Nature Protocols, 10(4), 548-557 (2015)
T Martin et al.
Chemical communications (Cambridge, England), (1)(1), 24-25 (2002-07-18)
Texture-related features of water intrusion in hydrophobised MCM-41 silicas render these materials especially suitable for energy dissipation in mechanical dampers.
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service