Skip to Content
Merck
All Photos(1)

Key Documents

236799

Supelco

Silica gel Inorganic Sorbent

high-purity grade (Davisil Grade 635), 60-100 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$124.00
1 KG
$625.00

$124.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$124.00
1 KG
$625.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
NB.21

$124.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Product Name

Silica gel, high-purity grade (Davisil Grade 635), pore size 60 Å, 60-100 mesh

grade

high-purity grade (Davisil Grade 635)

Quality Level

Assay

≥99%

form

powder or granules

manufacturer/tradename

Davisil®

technique(s)

LPLC: suitable

surface area

480 m2/g

matrix

Silica

matrix active group

silica

particle size

150-250 μm
60-100 mesh

pore size

0.75 cm3/g pore volume
60 Å

pH

6.5 (5% in slurry)

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is a rigid 3D network of colloidal silica.[1] It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density.[2] Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction[1]

Application

Silica gel was used to compare MCM-41 type mesoporous adsorbents.[3]

Legal Information

Davisil is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joonwoo Park et al.
The American journal of Chinese medicine, 48(8), 1841-1858 (2020-12-11)
Bisphenol A (BPA), which is known to be an endocrine-disrupting chemical (EDC), is associated not only with estrogen activity and reproductive toxicity but also with a variety of metabolic disorders. BPA affects glucose tolerance, cholesterol biosynthesis, and fatty acid synthesis.
Eilhann E Kwon et al.
Environmental research, 183, 109199-109199 (2020-02-07)
In this study, it is introduced a sustainable synthetic route of alkyl esters, considered value-added industrial chemicals and fuels, from volatile fatty acids (VFAs) that can potentially be generated from organic waste. In the presence of a porous carbon material
T Martin et al.
Chemical communications (Cambridge, England), (1)(1), 24-25 (2002-07-18)
Texture-related features of water intrusion in hydrophobised MCM-41 silicas render these materials especially suitable for energy dissipation in mechanical dampers.
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service