Skip to Content
Merck
All Photos(1)

Key Documents

214477

Supelco

Silica gel Inorganic Sorbent

high-purity grade (Davisil Grade 923), 100-200 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$116.00
250 G
$336.00
1 KG
$1,030.00

$116.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
50 G
$116.00
250 G
$336.00
1 KG
$1,030.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352309
PubChem Substance ID:
NACRES:
SB.52

$116.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Product Name

Silica gel, high-purity grade (Davisil Grade 923), pore size 30 Å, 100-200 mesh

grade

high-purity grade (Davisil Grade 923)

Quality Level

form

fine crystals

manufacturer/tradename

Davisil®

technique(s)

LPLC: suitable

surface area

430-530 m2/g

matrix

Silica

matrix active group

silica

particle size

100-200 mesh
75-150 μm

pore size

0.40 cm3/g pore volume
30 Å

bp

2230 °C

mp

>1600 °C

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silica gel is a rigid 3D network of colloidal silica.[1] It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation lresulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density.[2] Silica gel is classified as below:
a. Aquagel - pores are filled with water
b. Xerogel - by the process of evaporation, aqueous phase in the pores are removed
c. Aerogel - solvent removed by supercritical extraction[1]

Application

Silica gel was used to compare MCM-41 type mesoporous adsorbents.[3]

Footnote

Recommended Applications
Grade 923 meets ASTM D1319 specifications for hydrocarbon analysis

Legal Information

Davisil is a registered trademark of W.R. Grace & Co.-Conn.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Megan P Caputo et al.
Brain, behavior, and immunity, 81, 455-469 (2019-07-05)
Neonatal brain development can be disrupted by infection that results in microglial cell activation and neuroinflammation. Studies indicate that polyunsaturated fatty acids (PUFAs) and their metabolites can resolve inflammation. It is not known if dietary PUFA increases lipid metabolites in
T Martin et al.
Chemical communications (Cambridge, England), (1)(1), 24-25 (2002-07-18)
Texture-related features of water intrusion in hydrophobised MCM-41 silicas render these materials especially suitable for energy dissipation in mechanical dampers.
A simple method for production of pure silica from rice hull ash
Kalapathy, U., A. Proctor, and J. Shultz.
Bioresource Technology, 73, 257-262 (2000)
Silica gel supported zirconocene dichloride/methylalumoxane catalysts for ethylene polymerization: Effects of heterogenation on activity, polymer microstructure and product morphology.
Janiak, Christoph, and Bernhard Rieger.
Angew. Makromol. Chem., 215, 47-57 (1994)

Related Content

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service