Skip to Content
Merck
All Photos(1)

Documents

18370

Supelco

2,2,2-Trifluoroethanol

suitable for mass spectrometry (MS), LiChropur, ≥99.8% (GC)

Synonym(s):

Trifluoroethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH2OH
CAS Number:
Molecular Weight:
100.04
Beilstein:
1733203
EC Number:
MDL number:
UNSPSC Code:
41116105

vapor density

3.5 (vs air)

Quality Level

vapor pressure

70 mmHg ( 25 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

~896 °F

expl. lim.

42 %

manufacturer/tradename

LiChropur

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.390-1.392 g/mL at 20 °C
1.373 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤1 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

suitability

complies for LC-MS

SMILES string

OCC(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2

InChI key

RHQDFWAXVIIEBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

  • 2,2,2-Trifluoroethanol is commonly used as a mobile phase additive in liquid chromatography-mass spectrometry (LC-MS).

Features:
  • Extensive QC testing ensures the highest specifications.,·        
  • Optimized to improve ionization and resolution.
  • Extremely low levels of inorganic and organic impurities.      
  • Manufactured specifically for accurate and fast LC-MS.

Application

2,2,2-Trifluoroethanol may be used as a solvent in the development of a sample preparation method for the detection of high-mass proteins in tissue samples using matrix-assisted laser desorption/ionization mass spectrometry technique. It may also be used as a mobile phase modifier for the analysis and purification of alcohol-sensitive chiral compounds using supercritical fluid chromatography (SFC) technique.

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MALDI mass spectrometry imaging of proteins exceeding 30,000 daltons
Franck J, et al.
Medical Science Monitor : International Medical Journal of Experimental and Clinical Research, 16(9), BR293-BR299 (2010)
Analysis and purification of alcohol-sensitive chiral compounds using 2, 2, 2-trifluoroethanol as a modifier in supercritical fluid chromatography
Byrne N, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 875(1), BR293-BR299 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service