Skip to Content
Merck
All Photos(1)

Key Documents

91683

Supelco

2,2,2-Trifluoroethanol

analytical standard, for NMR spectroscopy

Synonym(s):

Trifluoroethyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$197.00
100 ML
$716.00

$197.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$197.00
100 ML
$716.00

About This Item

Linear Formula:
CF3CH2OH
CAS Number:
Molecular Weight:
100.04
Beilstein:
1733203
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

$197.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard
for NMR spectroscopy

Quality Level

vapor density

3.5 (vs air)

vapor pressure

70 mmHg ( 25 °C)

Assay

≥99.5% (GC)

autoignition temp.

~896 °F

expl. lim.

42 %

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.373 g/mL at 25 °C (lit.)

suitability

suitable for NMR (spectroscopy)

application(s)

environmental

format

neat

SMILES string

OCC(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2

InChI key

RHQDFWAXVIIEBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,2-Trifluoroethanol may be used as a co-solvent for the purification of certain alcohol sensitive compounds like boronic acid pinacol esters using supercritical fluid chromatography technique(SFC).[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Applications of 2, 2, 2 trifluoroethanol as a versatile co-solvent in supercritical fluid chromatography for purification of unstable boronate esters, enhancing throughput, reducing epimerization, and for additive free purifications
Asokan K, et al.
Journal of Chromatography A, 1531, 122-130 (2018)
Bei Feng et al.
Biomacromolecules, 13(12), 3917-3925 (2012-11-08)
In tissue engineering research, there has recently been considerable interest in using electrospun biomimetic nanofibers of hybrids, in particular, from natural and synthetic polymers for engineering different tissues. However, phase separation between a pair of much dissimilar polymers might give
A facile parallel synthesis of trifluoroethyl-substituted alkynes.
Cui-Bo Liu et al.
Angewandte Chemie (International ed. in English), 51(25), 6227-6230 (2012-05-09)
Catherine Pan et al.
Journal of proteome research, 13(11), 4535-4545 (2014-05-24)
Despite extensive research, an unmet need remains for protein biomarkers of Parkinson's disease (PD) in peripheral body fluids, especially blood, which is easily accessible clinically. The discovery of such biomarkers is challenging, however, due to the enormous complexity and huge
John P Hodkinson et al.
Rapid communications in mass spectrometry : RCM, 26(16), 1783-1792 (2012-07-11)
Amyloid formation is implicated in a number of human diseases. β(2)-Microglobulin (β(2)m) is the precursor protein in dialysis-related amyloidosis and it has been shown that partial, or more complete, unfolding is key to amyloid fibril formation in this pathology. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service