Skip to Content
Merck
All Photos(1)

Key Documents

73345

Supelco

Dibutylammonium acetate solution

0.5 M in H2O, LiChropur, suitable for ion pair chromatography

Synonym(s):

Dibutylamine acetate Concentrate

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
$601.00

$601.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
6 EA
$601.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)2NH · CH3COOH
CAS Number:
Molecular Weight:
189.30
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21

$601.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

form

solution

Quality Level

quality

LiChropur

packaging

pkg of 25 ML Volume containing 10 ML of product

concentration

0.5 M in H2O

technique(s)

LC/MS: suitable
ion pair chromatography: suitable

refractive index

n20/D 1.348

SMILES string

CC(O)=O.CCCCNCCCC

InChI

1S/C8H19N.C2H4O2/c1-3-5-7-9-8-6-4-2;1-2(3)4/h9H,3-8H2,1-2H3;1H3,(H,3,4)

InChI key

MQFIKAWTCOXAAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dibutylammonium acetate is an ion-pairing agent with quaternary ammonium ions. It generally helps in ionization of carbohydrate fragments providing a volatile solvent for the mass spectrometric technique.

Application

It has been used as buffer in mobile phase for separation and quantifying intracellular nucleotides from different microorganisms using ion pair reversed phase liquid chromatography electrospray ionization isotope dilution tandem mass spectrometry (IP–LC–ESI–ID–MS/MS) method.[1]

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)
concentrate in ampules for 6x1 l 0.005 M ready-to-use solution

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous quantification of free nucleotides in complex biological samples using ion pair reversed phase liquid chromatography isotope dilution tandem mass spectrometry.
Seifar, Reza M., et al.
Analytical Biochemistry, 388.2, 213-219 (2009)
Valentina Izzo et al.
Cell cycle (Georgetown, Tex.), 16(3), 271-279 (2017-01-07)
Phase II clinical trials indicate that the combination of cysteamine plus epigallocatechin gallate (EGCG) is effective against cystic fibrosis in patients bearing the most frequent etiological mutation (CFTRΔF508). Here, we investigated the interaction between both agents on cultured respiratory epithelia
Christian D Laourdakis et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 967, 127-133 (2014-08-05)
Targeted metabolite profiling has aided in the understanding of a variety of biological processes in the malaria parasite as well as in drug discovery. A fast and sensitive analytical method, based on ion-pairing reversed phase ultra-high performance liquid chromatography tandem

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service