Skip to Content
Merck
All Photos(1)

Key Documents

16237

Supelco

2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl acetate

pharmaceutical impurity standard

Synonym(s):

Aciclovir Related Compound A (USP), Aciclovir impurity A (PhEur)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% HPLC impurities
≤5% related solvents
≤5% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)OCCOCn1cnc2C(=O)NC(N)=Nc12

InChI

1S/C10H13N5O4/c1-6(16)19-3-2-18-5-15-4-12-7-8(15)13-10(11)14-9(7)17/h4H,2-3,5H2,1H3,(H3,11,13,14,17)

InChI key

DMHAXLGAKQREIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl acetate (impurity A) is the acetate form of acyclovir. It can be path indicative in the synthesis of acyclovir.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Huidobro et al.
Journal of pharmaceutical and biomedical analysis, 37(4), 687-694 (2005-03-31)
Acyclovir, guanine, and impurity A have been baseline separated with isocratic conditions at pH=3.0 and run time under 15 min by employing a SB-CN column from Agilent (150 mm x 4.6mm and 3.5 microm). Moreover, when run time was increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service