Skip to Content
Merck
All Photos(1)

Key Documents

254703

Sigma-Aldrich

2-Methoxybenzoyl chloride

97%

Synonym(s):

o-Anisoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$88.40
25 G
$175.00

$88.40


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$88.40
25 G
$175.00

About This Item

Linear Formula:
CH3OC6H4COCl
CAS Number:
Molecular Weight:
170.59
Beilstein:
607605
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$88.40


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.572 (lit.)

bp

128-129 °C/8 mmHg (lit.)

density

1.146 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

COc1ccccc1C(Cl)=O

InChI

1S/C8H7ClO2/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3

InChI key

RZNHSEZOLFEFGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methoxybenzoyl chloride has been used in preparation of:
  • 2-methoxybenzoyl-ferrocene Friedel Crafts reaction with ferrocene[1]
  • 2-methoxybenzoyl phosphate[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferrocenes. IV. Hydroxybenzoylferrocenes and Ethyl Ferrocenecarboxylates1, 2.
SCHAAF RL.
The Journal of Organic Chemistry, 27(1), 107-111 (1962)
P Paoli et al.
Experientia, 51(1), 57-62 (1995-01-15)
A new aromatic acyl phosphate, 2-methoxybenzoyl phosphate, has been synthesized. The compound shows an intrinsic fluorescence; it displays an intense emission band at 390 nm upon excitation in the near UV region. This band practically disappears after hydrolysis of the
Ataf Ali Altaf et al.
Dalton transactions (Cambridge, England : 2003), 47(34), 11769-11781 (2018-08-18)
Twenty-three compounds in two series of ferrocene-based anilides, with the general formula C5H5-Fe-C5H4-C6H4-NH-CO-C6H4-R (where R = H, F, Cl, CH3 and OCH3), have been successfully synthesized. The compounds were characterized by elemental analysis and FTIR, 1H NMR and 13C NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service