Skip to Content
Merck
All Photos(1)

Documents

119814

Sigma-Aldrich

Sodium 2,6-dichloroindophenolate hydrate

ACS reagent

Synonym(s):

2,6-Dichloro-N-(4-hydroxyphenyl)-1,4-benzoquinoneimine sodium salt, 2,6-Dichloroindophenol sodium salt hydrate, 2,6-Dichlorophenolindophenol sodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6Cl2NNaO2 · xH2O
CAS Number:
Molecular Weight:
290.08 (anhydrous basis)
Beilstein:
3641229
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

solid

loss

≤12.0% loss on drying

λmax

605 nm

application(s)

food and beverages

SMILES string

[Na+].[H]O[H].[O-]c1ccc(cc1)\N=C2/C=C(Cl)C(=O)C(Cl)=C2

InChI

1S/C12H7Cl2NO2.Na.H2O/c13-10-5-8(6-11(14)12(10)17)15-7-1-3-9(16)4-2-7;;/h1-6,16H;;1H2/q;+1;/p-1

InChI key

XHSOLXWCGCVQHE-UHFFFAOYSA-M

Related Categories

Application

Sodium 2,6-dichloroindophenolate hydrate can be used as a titration indicator to determine Vitamin C content of fruits.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivek K Vyas et al.
European journal of medicinal chemistry, 82, 385-393 (2014-06-16)
In continuation of our research for novel human dihydroorotate dehydrogenase (hDHODH) inhibitors, herein we reported design, synthesis and pharmacological evaluation of novel substituted quinoline-2-carboxamide derivatives. Human DHODH enzyme inhibition assay was used to screen the synthesized compounds as hDHODH inhibitors.
Dan Dong et al.
Bioscience, biotechnology, and biochemistry, 78(11), 1963-1970 (2014-07-24)
A chemolithoautotrophic arsenite-oxidizing bacterium, designated strain KGO-5, was isolated from arsenic-contaminated industrial soil. Strain KGO-5 was phylogenetically closely related with Sinorhizobium meliloti with 16S rRNA gene similarity of more than 99%, and oxidized 5 mM arsenite under autotrophic condition within
Yongsheng Chen et al.
Journal of agricultural and food chemistry, 63(1), 169-176 (2014-12-11)
Consumption of plant foods has been negatively associated with the risk of developing chronic diseases, which is partly attributed to their rich and diverse phytochemicals. To promote the rational and effective application of Adinandra tea (Adinandra Jack), a traditional Chinese
Justyna Sulej et al.
Applied biochemistry and biotechnology, 176(6), 1638-1658 (2015-05-25)
Cellobiose dehydrogenase (CDH), a secreted flavocytochrome produced by a number of wood-degrading fungi, was detected in the culture supernatant of a biotechnologically important strain of Cerrena unicolor grown in a modified cellulose-based liquid medium. The enzyme was purified as two
Fernando Dobrachinski et al.
Neurotoxicity research, 26(4), 317-330 (2014-03-13)
Acute stroke is a major risk for morbidity and mortality in aging population. Mitochondrion has been the focus of a wide stroke-related research. This study investigated if treatment or pre-treatment with diphenyl diselenide (PhSe)2 can prevent mitochondrial damage in cerebral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service