Merck
  • Characterization of Cellobiose Dehydrogenase from a Biotechnologically Important Cerrena unicolor Strain.
Characterization of Cellobiose Dehydrogenase from a Biotechnologically Important Cerrena unicolor Strain.
Applied biochemistry and biotechnology (2015-05-25)
Justyna Sulej, Grzegorz Janusz, Monika Osińska-Jaroszuk, Patrycja Rachubik, Andrzej Mazur, Iwona Komaniecka, Adam Choma, Jerzy Rogalski
ABSTRACT
Cellobiose dehydrogenase (CDH), a secreted flavocytochrome produced by a number of wood-degrading fungi, was detected in the culture supernatant of a biotechnologically important strain of Cerrena unicolor grown in a modified cellulose-based liquid medium. The enzyme was purified as two active fractions: CuCDH-FAD (flavin domain) (1.51-fold) with recovery of 8.35 % and CuCDH (flavo-heme enzyme) (21.21-fold) with recovery of 73.41 %. As CDH from other wood-rotting fungi, the intact form of cellobiose dehydrogenase of C. unicolor is a monomeric protein containing one flavin and one heme b with molecular mass 97 kDa and pI = 4.55. The enzyme is glycosylated (8.2 %) mainly with mannose and glucosamine residues. Moreover, the cellobiose dehydrogenase gene cdh1 and its corresponding cDNA from the fungus C. unicolor were isolated, cloned, and characterized. The 2316-bp full-length cDNA of cdh1 encoded a mature CDH protein containing 771 amino acids preceded by a signal peptide consisting of 18 amino acids. Moreover, both active fractions were characterized in terms of kinetics, temperature and pH optima, and antioxidant properties.
Sigma-Aldrich
2,6-Dichloroindophenol sodium salt hydrate, suitable for vitamin C determination, BioReagent
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium 2,6-dichloroindophenolate hydrate, ACS reagent
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate solution, BioUltra, ~3 M in H2O