Skip to Content
Merck
All Photos(1)

Documents

09709

Sigma-Aldrich

Ammonium phosphate monobasic

BioUltra, ≥99.5% (T)

Synonym(s):

mono-Ammonium phosphate, prim-Ammonium phosphate, Ammonium dihydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4H2PO4
CAS Number:
Molecular Weight:
115.03
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥99.5% (T)

impurities

insoluble matter, passes filter test

pH

4.0-5.0 (25 °C, 0.1 M in H2O)

mp

190 °C (dec.) (lit.)

solubility

H2O: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.059
λ: 280 nm Amax: ≤0.050

SMILES string

N.OP(O)(O)=O

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium phosphate monobasic can be used to study optimization reagents, protein structural analysis, proteomics, x-ray crystallography and biological buffers. Ammonium phosphate monobasic has been used to develop a simple high-performance liquid chromatographic method for determining N1-alkylnicotinamides, including C1C5 alkyl derivatives, in urine. Ammonium phosphate monobasic has also been used to develop a high-performance liquid chromatographic assay coupled with UV detection (254 nm) for the determination of midazolam and two of its hydroxylated metabolites in human plasma.

Other Notes

Component of culture media e.g. DEV SIMMONS Citrate agar. Acetate agar acc. to Trabulsi and Ewing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.R. Trabulsi et al.
Publ. Hlth. Lab., 20, 137-137 (1962)
T Hirayama et al.
Analytical biochemistry, 147(1), 108-113 (1985-05-15)
A simple high-performance liquid chromatographic method has been developed for determining N1-alkylnicotinamides, including C1-C5 alkyl derivatives, in urine. N1-Alkylnicotinamides were reacted with acetophenone in strong alkali medium at 0 degrees C and then formic acid was added. The reaction mixture
María J Nozal et al.
Journal of pharmaceutical and biomedical analysis, 30(2), 285-291 (2002-08-23)
A high performance liquid chromatographic (HPLC) method for the assay of sumatriptan succinate residues in swabs collected from manufacturing equipment surfaces was developed and validated in order to control a cleaning procedure. The swabbing procedure using two cotton swabs moistened
Anelise Rodolfo Ferreira Pieralini et al.
The Journal of prosthetic dentistry, 108(1), 51-57 (2012-07-07)
Coatings of zirconite, Y(2)O(3) or ZrO(2) on wax patterns before investing in phosphate-bonded investments have been recommended to reduce the reaction layer in titanium castings, but they are not easily obtainable. Spinel-based investments are relatively stable with molten titanium and
Catarina Barbosa et al.
International journal of food microbiology, 160(2), 87-93 (2012-11-28)
Sulphur-containing amino acids, cysteine and methionine, are generally found in very low concentrations in grape-juice. The objective of this study was to identify the effects of methionine on aroma compounds formation. Nitrogen source effects on growth, fermentative behaviour and aroma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service