Skip to Content
Merck
All Photos(2)

Key Documents

379980

Sigma-Aldrich

Ammonium phosphate dibasic

≥99.99% trace metals basis

Synonym(s):

Ammonium hydrogenphosphate, Diammonium hydrogenphosphate, di-Ammonium hydrogenphosphate (sec)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$143.00
100 G
$489.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$143.00
100 G
$489.00

About This Item

Linear Formula:
(NH4)2HPO4
CAS Number:
Molecular Weight:
132.06
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

$143.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

for analytical purposes

Assay

≥99.99% trace metals basis

form

crystalline

impurities

≤100.0 ppm Trace Metal Analysis

mp

155 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

N.N.OP(O)(O)=O

InChI

1S/2H3N.H3O4P/c;;1-5(2,3)4/h2*1H3;(H3,1,2,3,4)

InChI key

MNNHAPBLZZVQHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ammonium phosphate dibasic is widely used as a starting material to fabricate cathodes for Li-ion batteries.[1] It can also be used to remove residual lithium from LiNiO2-based cathode materials.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced electrochemical properties of a LiNiO2-based cathode material by removing lithium residues with (NH4)2HPO4
Xunhui Xiong, et al.
Journal of Materials Chemistry, 2, 11691-11696 (2014)
Emulsion drying synthesis of olivine LiFePO4/C composite and its electrochemical properties as lithium intercalation material
Seung-Taek Myung, et al
Electrochimica Acta, 49, 4213-4222 (2004)
Janet Sze-Jing Cheung et al.
International journal of hygiene and environmental health, 230, 113623-113623 (2020-09-16)
There are concerns in Yellowknife, Northwest Territories, Canada, about arsenic exposure due to past mining operations, particularly the former Giant Mine. The objective of this study was to characterize the risk of arsenic exposure and associated risk factors among the
Martin P Pothier et al.
Frontiers in microbiology, 9, 2310-2310 (2018-10-20)
Despite its high toxicity and widespread occurrence in many parts of the world, arsenic (As) concentrations in decentralized water supplies such as domestic wells remain often unquantified. One limitation to effective monitoring is the high cost and lack of portability
Everest Akaraonye et al.
Biotechnology journal, 7(2), 293-303 (2011-12-08)
The main hindrance in the use of polyhydroxyalkanoates (PHAs) as a replacement for existing petroleum-based plastics is their high production cost. The carbon source accounts for 50% of the cost for PHA production. Thus, increasing the yield and productivity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service