Skip to Content
Merck
All Photos(1)

Key Documents

1.01440

Supelco

Ammonium dihydrogen phosphate

99.99 Suprapur®

Synonym(s):

Ammonium dihydrogen phosphate, Ammonium biphosphate, Ammonium phosphate monobasic, Primary ammonium phosphate, Monoammonium orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)H2PO4
CAS Number:
Molecular Weight:
115.03
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-764-5
NACRES:
NB.21
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

vapor pressure

0.066 hPa ( 125 °C)

Assay

99.99%

form

solid

potency

5750 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

storage condition

(Tightly closed. Dry.)

technique(s)

HPLC: suitable using : suitable

color

colorless

pH

3.8-4.4 (20 °C, 50 g/L in H2O)

mp

190 °C

solubility

368 g/L

density

1.80 g/cm3 at 20 °C

bulk density

800‑1100 kg/m3

anion traces

nitrate (NO3-): ≤5 ppm
sulfate (SO42-): ≤50 ppm

cation traces

Ba: ≤2.0 ppm
Ca: ≤0.10 ppm
Cd: ≤0.010 ppm
Ce: ≤0.01 ppm
Co: ≤0.010 ppm
Cr: ≤0.010 ppm
Cs: ≤50 ppm
Cu: ≤0.010 ppm
Fe: ≤0.10 ppm
Hg: ≤0.05 ppm
K: ≤10 ppm
La: ≤0.010 ppm
Li: ≤1.0 ppm
Mg: ≤0.10 ppm
Mn: ≤-0.010 ppm
Na: ≤10 ppm
Ni: ≤0.010 ppm
Pb: ≤0.010 ppm
Rb: ≤10 ppm
Sm: ≤0.010 ppm
Sr: ≤0.10 ppm
Tl: ≤0.01 ppm
Y: ≤0.010 ppm
Yb: ≤0.010 ppm
Zn: ≤0.010 ppm

storage temp.

2-30°C

SMILES string

[NH4+].OP(=O)(O)[O-]

InChI

1S/H3N.H3O4P/c;1-5(2,3)4/h1H3;(H3,1,2,3,4)

InChI key

LFVGISIMTYGQHF-UHFFFAOYSA-N

Application


  • Development and Characterization of Bio-Based Formaldehyde Free Sucrose-Based Adhesive for Fabrication of Plywood.: This study presents a bio-based adhesive using sucrose and ammonium dihydrogen phosphate, offering a formaldehyde-free solution for plywood fabrication, enhancing environmental sustainability (Liu et al., 2024).

  • Significantly Tunable Foaming Behavior of Blowing Agent for the Polyethylene Foam Resin with a Unique Designed Blowing Agent System.: This paper investigates a new blowing agent system incorporating ammonium dihydrogen phosphate, which allows significant tunability in the foaming behavior of polyethylene foam resin, highlighting applications in material science (Chen & Huang, 2024).

  • Facile preparation of high-performance plywood adhesive from gelatinized corn starch crosslinked with ammonium dihydrogen phosphate.: This article details the preparation of a high-performance plywood adhesive using gelatinized corn starch crosslinked with ammonium dihydrogen phosphate, offering an eco-friendly alternative to traditional adhesives (Jia et al., 2024).

Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (acidimetric): ≥ 99 %
Nitrate (NO₃): ≤ 5 ppm
Sulfate (SO₄): ≤ 50 ppm
As (Arsenic): ≤ 0.2 ppm
Ba (Barium): ≤ 2.0 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.01 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cs (Cesium): ≤ 50 ppm
Cu (Copper): ≤ 0.010 ppm
Fe (Iron): ≤ 0.10 ppm
Hg (Mercury): ≤ 0.05 ppm
K (Potassium): ≤ 10 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 1.0 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.010 ppm
Na (Sodium): ≤ 10 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.010 ppm
Rb (Rubidium): ≤ 10 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service