Skip to Content
Merck
All Photos(1)

Key Documents

07552

Supelco

Dihydrocoumarin

analytical standard

Synonym(s):

3,4-Dihydro-1-benzopyran-2-one, Benzodihydropyrone, Hydrocoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O2
CAS Number:
Molecular Weight:
148.16
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.555-1.559
n20/D 1.556 (lit.)

bp

272 °C (lit.)

mp

24-25 °C (lit.)

density

1.169 g/mL at 25 °C (lit.)

SMILES string

O=C1CCc2ccccc2O1

InChI

1S/C9H8O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-4H,5-6H2

InChI key

VMUXSMXIQBNMGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dihydrocoumarin is a naturally occurring compound found in Melilotus officinalis (sweet clover) species. It is predominantly used as a flavor ingredient in the food industry[1][2] and reported to exhibit immunomodulatory, anti-inflammatory, and antioxidant properties.[3]

Application

Dihydrocoumarin may be used as an analytical reference standard for the determination of the analyte in guaco extracts and pharmaceutical preparations, and various plant extracts by chromatography-based techniques[3] and capillary electrophoresis.[4][5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>212.0 °F - closed cup

Flash Point(C)

> 100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biocatalytic Synthesis of Natural Dihydrocoumarin by Microbial Reduction of Coumarin
Serra S, et al.
Catalysts, 9(8), 665-665 (2019)
Simultaneous determination of coumarin, o-coumaric acid, dihydrocoumarin and syringaldehyde in guaco extracts and pharmaceutical preparations by HPLC-DAD
Gasparetto JC, et al.
Pharmaceutica Analytica Acta, 2(145), 2-2 (2011)
Rapid determination of coumarins in plants by capillary electrophoresis
Kubrak T
Analytical Letters, 48(18), 2819-2832 (2015)
The flavoring agent dihydrocoumarin reverses epigenetic silencing and inhibits sirtuin deacetylases
Olaharski AJ, et al.
PLoS Genetics, 1(6) (2005)
Determination of coumarins from Chrysanthemum segetum L. by capillary electrophoresis
Ochocka RJ, et al.
Journal of Chromatography A, 709(1), 197-202 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service