Skip to Content
Merck
Dihydrocoumarin ≥99%, FCC, FG

Dihydrocoumarin

Dihydrocoumarin ≥99%, FCC, FG
Synonym(s):
3,4-Dihydro-1-benzopyran-2-one, Benzodihydropyrone, Hydrocoumarin
Empirical Formula (Hill Notation):
C9H8O2
CAS Number:
Molecular Weight:
148.16
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing