Skip to Content
Merck
All Photos(1)

Key Documents

W229105

Sigma-Aldrich

Cinnamon bark oil

FCC, FG

Synonym(s):

Cassia oil, Cinnamomum

Sign Into View Organizational & Contract Pricing

Select a Size

W229105-SAMPLE-K
$59.00
250 G
$437.00
1 KG
$1,000.00

$59.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W229105-SAMPLE-K
$59.00
250 G
$437.00
1 KG
$1,000.00

About This Item

CAS Number:
FEMA Number:
2291
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
Cinnamonum zeylanicum Blume
food allergen:
no known allergens

$59.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk OrderRequest more information

biological source

Cinnamonum zeylanicum Blume

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

optical activity

[α]20/D −1.0°, neat

origin

Sri Lanka origin

refractive index

n20/D 1.581

density

1.025 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cinnamon; spicy; sweet

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Inhibitory Properties of Cinnamon Bark Oil against Postharvest Pathogen Penicillium digitatum In Vitro.: Research highlights the antifungal properties of cinnamon bark oil, making it a potential natural preservative for postharvest fruit protection (Zhou et al., 2024).


Preparation Note

Extraction method: steam distillation

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service