Skip to Content
Merck
All Photos(2)

Key Documents

05757

Supelco

2′,6′-Dihydroxyacetophenone

matrix substance for MALDI-MS, ≥99.5% (HPLC), Ultra pure

Synonym(s):

2-Acetyl-1,3-dihydroxybenzene, 2-Acetylresorcinol, DHAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3COCH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
1366061
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

grade

matrix substance for MALDI-MS

Quality Level

Assay

≥99.5% (HPLC)

form

powder

mp

156-158 °C (lit.)

solubility

dioxane: 50 mg/mL, clear

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

CC(=O)c1c(O)cccc1O

InChI

1S/C8H8O3/c1-5(9)8-6(10)3-2-4-7(8)11/h2-4,10-11H,1H3

InChI key

YPTJKHVBDCRKNF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen D Inutan et al.
Molecular & cellular proteomics : MCP, 10(2), M110-M110 (2010-09-22)
Laserspray ionization (LSI) mass spectrometry (MS) allows, for the first time, the analysis of proteins directly from tissue using high performance atmospheric pressure ionization mass spectrometers. Several abundant and numerous lower abundant protein ions with molecular masses up to ∼20,000
Junjie Hou et al.
Journal of biomedicine & biotechnology, 2010, 759690-759690 (2010-03-27)
Selecting an appropriate matrix solution is one of the most effective means of increasing the ionization efficiency of phosphopeptides in matrix-assisted laser-desorption/ionization time-of-flight mass spectrometry (MALDI-TOF-MS). In this study, we systematically assessed matrix combinations of 2, 6-dihydroxyacetophenone (DHAP) and diammonium
MALDI mass spectrometry in the solution of some forensic problems.
R Seraglia et al.
Forensic science international, 146 Suppl, S83-S85 (2005-01-11)
Ana Maria Balan et al.
European journal of medicinal chemistry, 44(5), 2275-2279 (2008-08-02)
Herein we report a feasible study concerning syntheses, structure and antimicrobial activity of some new diazinium salts with dihydroxyacetophenone skeleton. A fast, general, environmentally friendly, and facile method for preparation of diazinium salts under microwave and ultrasounds irradiation is presented.
Erica L McJimpsey et al.
Journal of the American Society for Mass Spectrometry, 19(3), 315-324 (2007-12-25)
The Bioaerosol Mass Spectrometry (BAMS) system was developed for the real-time detection and identification of biological aerosols using laser desorption ionization. Greater differentiation of particle types is desired; consequently MALDI techniques are being investigated. The small sample size ( approximately

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service