Skip to Content
Merck
All Photos(1)

Documents

04626

Supelco

Hop-22(29)-ene solution

0.1 mg/mL in isooctane, analytical standard

Synonym(s):

Diploptene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50
CAS Number:
Molecular Weight:
410.72
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

0.1 mg/mL in isooctane
100 μg/mL in isooctane

application(s)

food and beverages

format

single component solution

storage temp.

−20°C

SMILES string

[H][C@@]12CC[C@]3(C)[C@]([H])(CC[C@]4([H])[C@@]5(C)CCCC(C)(C)[C@]5([H])CC[C@@]34C)[C@@]1(C)CC[C@@H]2C(C)=C

InChI

1S/C30H50/c1-20(2)21-12-17-27(5)22(21)13-18-29(7)24(27)10-11-25-28(6)16-9-15-26(3,4)23(28)14-19-30(25,29)8/h21-25H,1,9-19H2,2-8H3/t21-,22+,23+,24-,25-,27+,28+,29-,30-/m1/s1

InChI key

HHXYJYBYNZMZKX-PYQRSULMSA-N

General description

Certan Vial

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dirk J Reinert et al.
Chemistry & biology, 11(1), 121-126 (2004-04-29)
The membrane protein squalene-hopene cyclase was cocrystallized with 2-azasqualene and analyzed by X-ray diffraction to 2.13 A resolution. The conformation of this close analog was clearly established, and it agreed with the common textbook presentation. The bound squalene undergoes only
M Vyssotski et al.
Lipids, 46(12), 1155-1161 (2011-08-02)
A Gram-negative, aerobic, pink-pigmented, rod-shaped bacterium Chthonomonas calidirosea (strain T49(T)) with an optimal temperature for growth of 68 °C, isolated from soil samples from Hell's Gate in the Tikitere geothermal system (New Zealand), was the first cultivated bacterium of the novel
B Andes Hess et al.
Organic letters, 6(11), 1717-1720 (2004-05-21)
The cyclization of the A-B rings of squalene to hopene is studied computationally (DFT). A transition structure is found for a concerted, asynchronous pathway for the formation of chair-chair decalin carbocation. The computationally derived conformer leading to this asynchronous transition
Tsutomu Sato et al.
Bioscience, biotechnology, and biochemistry, 68(3), 728-738 (2004-04-02)
To provide insight into the catalytic mechanism for the final deprotonation reaction of squalene-hopene cyclase (SHC) from Alicyclobacillus acidocaldarius, mutagenesis experiments were conducted for the following ten residues: Thr41, Glu45, Glu93, Arg127, Trp133, Gln262, Pro263, Tyr267, Phe434 and Phe437. An
H Schulenberg-Schell et al.
Analytical biochemistry, 181(1), 120-124 (1989-08-15)
A rapid and sensitive method for determination of various hopanoids in microorganisms is described. Tetrahydroxybacteriohopane (THBH), THBH-ether and -glycoside were acetylated with acetanhydride/pyridine and were separated on a C18 reversed-phase HPLC column with a gradient of acetonitrile in methanol and

Articles

We provide high-quality, relevant hopane reference standards to be used as biomarkers for petrochemical oil field remediation and spill analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service