Skip to Content
Merck
All Photos(2)

Key Documents

G9391

Sigma-Aldrich

Bovine Gelatin

from bovine skin, Type B, powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$57.30
500 G
$348.00

$57.30


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$57.30
500 G
$348.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.75

$57.30


Estimated to ship onMarch 29, 2025


Request a Bulk Order

Product Name

Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture

biological source

bovine skin

Quality Level

sterility

sterile

type

Type B

product line

BioReagent

form

powder

packaging

pkg of 100 g
pkg of 500 g

technique(s)

cell culture | mammalian: suitable

gel strength

~225 g Bloom

solubility

H2O: soluble 50 mg/mL, hazy to strongly hazy, faintly yellow to yellow

shipped in

ambient

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue. Gelatin is a hydrocolloid and is rich in glycine, proline and hydroxyproline, which impart structural stability. It is synthesized from the alkaline digestion of collagen from bovine skin and is referred as type B. It has wide applications in food industry.[1] Gelatin takes up random coil structure after digestion from the triple helical collagen.[2] The most common source for industrial production of gelatin is slaughter byproducts. The type A gelatin from porcine and type B differ in their isoelectric pH. The N-terminal sequence of bovine gelatin is unique for its identification. It has gelling property and displays surface behaviour for use in foams and adhesions.[3]

Application

Gelatin has been used:
  • in coating cell culture to improve attachment of cells,[4]
  • in PCR to stabilize Taq DNA, as a blocking reagent in Western blotting, ELISA, and immunochemistry,[5]
  • as a component of media for species differentiation in bacteriology
  • as a biocompatible polymer
  • as a delivery vehicle for the release of active biomolecules[4]
  • in the generation of scaffolds for tissue engineering applications.[6]
  • to study long-chain fatty acid-induced changes in gene expression in neonatal cardiac monocytes
  • to study to test mobilization of capillary endothelium in-vitro induced by effectors of angiogenesis in vivo
This product is recommended as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/mL. The optimal concentration depends on cell type and the application and research objectives.

Biochem/physiol Actions

Gelatin is commonly used in place of collagen due to its reduced immune antigenicity and cost-effectiveness. This macromolecule contains arginine-glycine-aspartic acid (RGD) sequences, known to promote osteoblast adhesion and enhance osteointegration. Its similar properties make gelatin a favorable substitute for collagen in many applications. Gelatin finds extensive use in enhancing cell attachment across various cell types. Moreover, it serves as a biocompatible polymer with applications in delivering bioactive molecules and creating scaffolds for tissue engineering purposes. This versatile material contributes to improved cell adhesion, controlled release of bioactive substances, and the development of tissue-engineered scaffolds.[7]

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Preparation Note

This product is derived from bovine skin. Gelatin is soluble in hot than in cold water. It is practically insoluble in most organic solvents such as alcohol, chloroform, carbon disulfide, carbon tetrachloride, ether, benzene, acetone, and oils. The Bloom number, determined by the Bloom gelometer, is an indication of the strength of a gel formed from a solution of the known concentration. The Bloom number is proportional to the average molecular mass. Bloom numbers of porcine skin Gelatin vary from 90 to 300 g.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tao Jiang et al.
Scientific reports, 7(1), 4575-4575 (2017-07-06)
Human tumour progression is a dynamic process involving diverse biological and biochemical events such as genetic mutation and selection in addition to physical, chemical, and mechanical events occurring between cells and the tumour microenvironment. Using 3D bioprinting we have developed
bFGF-containing electrospun gelatin scaffolds with controlled nano-architectural features for directed angiogenesis
Montero RB, et al.
Acta Biomaterialia, 8(5), 1778-1791 (2012)
A microparticle approach to morphogen delivery within pluripotent stem cell aggregates
Bratt-Leal AM, et al.
Biomaterials, 34(30), 7227-7235 (2013)
Kathryn E Drzewiecki et al.
Biophysical journal, 111(11), 2377-2386 (2016-12-08)
Type-I collagen assembles in a stepwise, hierarchic fashion from the folding of the triple helix to the assembly of fibrils into fibers. The mature assembled fibers are crucial for tissue structure and mechanics, cell interactions, and other functions in vivo. Although triple
Investigation and Application of HA Composite Coating on the Ti Alloy
Qiang W, et al.
Journal of Separation Science (2013)

Articles

The extracellular matrix (ECM) and its attachment factor components are discussed in this article in relation to their function in structural biology and their availability for in vitro applications.

The extracellular matrix (ECM) is secreted by cells and surrounds them in tissues.

Cancer stem cell media, spheroid plates and cancer stem cell markers to culture and characterize CSC populations.

Protocols

This gelatin coating protocol for cell culture applications offers information regarding types of gelatin, surface coverage concentration, and tips for optimization.

Questions

  1. What is the molecular weight of this Gelatin?

    1 answer
    1. The molecular weight of this product has not been evaluated, However, the molecular weight is closely related to the Bloom Number. The estimated ranges are as follows:
      Bloom Number Average Molecular Mass (Da)
      50−125 (Low Bloom) 20,000−25,000
      175−225 (Medium Bloom) 40,000−50,000
      225−325 (High Bloom) 50,000−100,000

      Please see the link below to review the general datasheet for Gelatin:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/333/625/g9382pis.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service